
Magnesium ionophore IV

Synonym(s):
ETH 7025, N,N′,N′′-Tris[3-(heptylmethylamino)-3-oxopropionyl]-8,8′-iminodioctylamine
Empirical Formula (Hill Notation):
C49H94N6O6
CAS Number:
Molecular Weight:
863.31
Products
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing