
Lipopolysaccharides from Salmonella typhosa

Synonym(s):
LPS
Products
Product Category
Brand
Biological Source
Color
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing