
Levorphanol tartrate

Synonym(s):
Levorphanol (+)-tartrate salt dihydrate, L-3-Hydroxy-N-methylmorphinan (+)-tartrate salt dihydrate
Empirical Formula (Hill Notation):
C17H23NO · C4H6O6 · 2H2O
CAS Number:
Molecular Weight:
443.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing