
Lectin from Ulex europaeus (gorse, furze)

Synonym(s):
Ulex europaeus agglutinin, UEA-I
Products
Brand
Biological Source
Physical Form
Chemical Composition
Conjugate
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing