Skip to Content
Merck
Irisflorentin European Pharmacopoeia (EP) Reference Standard

Irisflorentin

Irisflorentin European Pharmacopoeia (EP) Reference Standard
Synonym(s):
9-Methoxy-7-(3,4,5-trimethoxyphenyl)-8H-1,3-dioxolo[4,5-g][1]benzopyran-8-one
Empirical Formula (Hill Notation):
C20H18O8
CAS Number:
Molecular Weight:
386.35

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2