
Hyperoside

Synonym(s):
Quercetin 3-D-galactoside, 3,3′,4′,5,7-Pentahydroxyflavone 3-D-galactoside, Hyperin, Hyperoside
Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing