
Hydroxyzine hydrochloride

Synonym(s):
Hydroxyzine dihydrochloride
Empirical Formula (Hill Notation):
C21H27ClN2O2 · 2HCl
CAS Number:
Molecular Weight:
447.83
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade