
Hexylresorcinol

Synonym(s):
4-Hexylresorcinol, 4-Hexyl-1,3-dihydroxybenzene
Linear Formula:
CH3(CH2)5C6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
194.27
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing