
Guaifenesin Impurity D

Synonym(s):
1,3-Bis(2-methoxyphenoxy)-2-propanol, 1,3-Bis(2-methoxyphenoxy)propan-2-ol, NSC 142122
Empirical Formula (Hill Notation):
C17H20O5
CAS Number:
Molecular Weight:
304.34
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing