
Gram’s stain solution (III)

Synonym(s):
Basic Fuchsin, Basic Violet 14, Fuchsin basic, Magenta™, Rosaniline
Empirical Formula (Hill Notation):
C20H20ClN3
CAS Number:
Molecular Weight:
337.85
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing