
Glycerol monocaprylate

Empirical Formula (Hill Notation):
C11H22O4
Molecular Weight:
218.29
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing