
Glucose-6-phosphate

Synonym(s):
D-Glucose 6-phosphate disodium salt hydrate, G-6-P-Na2, Robison ester
Empirical Formula (Hill Notation):
C6H11Na2O9P · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
Products
Formula Weight
Manufacturer
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing