
Fluphenazine hydrochloride

Synonym(s):
Fluphenazine dihydrochloride, 4-[3-[2-(Trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol dihydrochloride
Empirical Formula (Hill Notation):
C22H26F3N3OS · 2HCl
CAS Number:
Molecular Weight:
510.44
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing