
Flecainide impurity B

Synonym(s):
2-(Aminomethyl)piperidine, (RS)-1-(Piperidin-2-yl)methanamine, 2-Piperidinemethanamine
Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing