
DMT-locT Phosphoramidite

Synonym(s):
DMT-locT Phosphoramidite
Empirical Formula (Hill Notation):
C41H49N4O9P
CAS Number:
Molecular Weight:
772.82
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing