
D-Glucose-2-d

Synonym(s):
Dextrose-2-d
Linear Formula:
HOCH2[CHOH]3CDOHCHO
CAS Number:
Molecular Weight:
181.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing