
Dehydro-L-(+)-ascorbic acid dimer

Synonym(s):
Bis(dehydro-L-ascorbic acid), Bis-DHA, DHA
Empirical Formula (Hill Notation):
C12H12O12
CAS Number:
Molecular Weight:
348.22
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing