
Clotrimazole impurity E

Synonym(s):
2-Chlorobenzophenone
Linear Formula:
ClC6H4COC6H5
CAS Number:
Molecular Weight:
216.66
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing