
Clear glass 1.2 mL crimp top vial, flat bottom, 8 x 40 mm

Synonym(s):
Vials for Waters® 96-position carousel, 8 x 40 mm
Products
Product Category
Brand
Manufacturer
Diameter (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing