
Candesartan cilexetil Related Compound A

Synonym(s):
(Ethyl 1-{[2′-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl}-2-ethoxybenzimidazole-7-carboxylate)
Empirical Formula (Hill Notation):
C26H24N6O3
CAS Number:
Molecular Weight:
468.51
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2