
Bis(triphenylphosphine)ruthenium(II) dicarbonyl chloride

Synonym(s):
Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II), Ruthenium(II)bis(triphenylphosphine)dichlorocarbonyl
Linear Formula:
[(C6H5)3P]2Ru(CO)2Cl2
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing