
Benzathine phenoxymethylpenicillin for system suitability

Synonym(s):
Benzathine phenoxymethylpenicillin
Empirical Formula (Hill Notation):
C16H2N2 · 2C16H18N2O5S
CAS Number:
Molecular Weight:
922.98
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing