
Anti-c-Myc−Peroxidase antibody produced in rabbit

Synonym(s):
Anti-c-Myc antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing