
Anti-Chicken IgY (IgG) (whole molecule)−FITC antibody produced in rabbit

Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing