
Anacardic acid

Synonym(s):
2-Hydroxy-6-pentadecylbenzoic acid, 22:0-Anacardic acid, 6-Pentadecylsalicylic acid, Anacardic Acid
Empirical Formula (Hill Notation):
C22H36O3
CAS Number:
Molecular Weight:
348.52
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment