
Amiloride impurity A

Synonym(s):
Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
Empirical Formula (Hill Notation):
C6H7ClN4O2
CAS Number:
Molecular Weight:
202.60
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing