
Albuterol Related Compound B

Synonym(s):
2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone, Salbutamon
Empirical Formula (Hill Notation):
C13H19NO3
CAS Number:
Molecular Weight:
237.29
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing