
Albendazole

Synonym(s):
Methyl 5-(propylthio)-2-benzimidazolecarbamate
Empirical Formula (Hill Notation):
C12H15N3O2S
CAS Number:
Molecular Weight:
265.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing