
Acetaldehyde-2,4-DNPH solution

Synonym(s):
Acetaldehyde-2,4-dinitrophenylhydrazone solution
Empirical Formula (Hill Notation):
C8H8N4O4
CAS Number:
Molecular Weight:
224.17
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2