
Acebutolol impurity B

Synonym(s):
N-{3-Acetyl-4-{(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy}phenyl}acetamide, Diacetolol
Empirical Formula (Hill Notation):
C16H24N2O4
CAS Number:
Molecular Weight:
308.37
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing