
5-Bromo-3-fluoropicolinonitrile

Empirical Formula (Hill Notation):
C6H2BrFN2
CAS Number:
Molecular Weight:
201.00
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing