
2-Trifluoromethylthioxanthone

Synonym(s):
2-(Trifluoromethyl)thioxanthen-9-one
Linear Formula:
C13H7SOCF3
CAS Number:
Molecular Weight:
280.26
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing