
1-Fluoronaphthalene

Empirical Formula (Hill Notation):
C10H7F
CAS Number:
Molecular Weight:
146.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2