Skip to Content
Merck
All Photos(1)

Key Documents

SML1210

Sigma-Aldrich

Almotriptan malate

≥98% (HPLC)

Synonym(s):

3-[2-(Dimethylamino)ethyl]-5-(pyrrolidin-1-ylsulfonylmethyl )-1H-indole malate, PNU 180638

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25N3O2S · C4H6O5
CAS Number:
Molecular Weight:
469.55
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

−20°C

InChI

1S/C17H25N3O2S.C4H6O5/c1-19(2)10-7-15-12-18-17-6-5-14(11-16(15)17)13-23(21,22)20-8-3-4-9-20;5-2(4(8)9)1-3(6)7/h5-6,11-12,18H,3-4,7-10,13H2,1-2H3;2,5H,1H2,(H,6,7)(H,8,9)

InChI key

QHATUKWEVNMHRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Almotriptan at an oral dose of 12.5 mg, is recommended to be safe for treating migraine.
Almotriptan is a serotonin 5HT-1B/1D-receptor agonist; antimigraine.

Biochem/physiol Actions

Almotriptan is a serotonin 5HT-1B/1D-receptor agonist used to treat migraine. Almotriptan has low nanomolar affinity for the 5-HT(1B) and 5-HT(1D) receptors while affinity for 5-HT receptors other than 5-HT(1B/1D) is substantially lower. Affinity for 5-HT(7) and 5-HT(1A) receptors was approximately 40 and 60 times lower than that for 5-HT(1B/1D) receptors, respectively.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dose finding, placebo-controlled study of oral almotriptan in the acute treatment of migraine
Dahlof C, et al.
Neurology, 57(10), 1811-1817 (2001)
Kremena Saracheva et al.
Acta pharmaceutica (Zagreb, Croatia), 70(2), 239-247 (2020-01-20)
The introduction of the second generation triptans in clinical and experimental practice was a major progress in the pharmacotherapy of migraine. Frovatriptan is a second generation triptan with strong 5-HT1B/1D serotonergic agonism and low 5-HT1A/7 receptor affinity, while almotriptan possesses
Long?term Efficacy and Safety of Oral Almotriptan: Interim Analysis of a 1?Year Open Study
Cabarrocas X, et al.
Headache, 41(1), 57-62 (2001)
Barbora Vyhlídalová et al.
International journal of molecular sciences, 21(8) (2020-04-23)
The efforts for therapeutic targeting of the aryl hydrocarbon receptor (AhR) have emerged in recent years. We investigated the effects of available antimigraine triptan drugs, having an indole core in their structure, on AhR signaling in human hepatic and intestinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service