Skip to Content
Merck
All Photos(1)

Key Documents

225M-1

Sigma-Aldrich

BCA-225 (Cu-18) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

Cu-18, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (225M-14)
vial of 0.5 mL concentrate (225M-15)
bottle of 1.0 mL predilute (225M-17)
vial of 1.0 mL concentrate (225M-16)
bottle of 7.0 mL predilute (225M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

breast

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-BCA-225 antibody recognizes a human breast carcinoma associated glycoprotein BCA-225 (220-225kD). This protein differs in size and distribution from other breast carcinoma antigens. Unlike other antibodies against breast carcinoma antigens, this antibody does not react with benign or malignant colonic, stomach, prostate, liver, pancreas, thyroid, or parotid tissues. Adenocarcinomas of the lung, ovary and endometrium also stain with this antibody.

Quality


IVD

IVD

IVD

RUO

Linkage

BCA-225 Positive Control Slides, Product No. 225S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T S Loy et al.
American journal of clinical pathology, 96(3), 326-329 (1991-09-01)
BCA-225 is a glycoprotein identified in human breast carcinoma cells that has been reported to show a restricted distribution in other human tissues. To further define the presence of BCA-225 in human carcinomas, the authors performed an immunohistochemical study, applying
Monoclonal Antibodies and Breast Cancer (1985)
R Mesa-Tejada et al.
The American journal of pathology, 130(2), 305-314 (1988-02-01)
A glycoprotein, BCA-225 (Mr 225,000-250,000), has been identified in cells and spent medium of clone 11 T47D breast carcinoma cells by three murine monoclonal antibodies, CU18, CU26, and CU46. The antigen was localized in paraffin sections of 167/178 (94%) Bouin's-fixed
C K Ma et al.
American journal of clinical pathology, 99(5), 551-557 (1993-05-01)
Distinguishing primary hepatocellular carcinoma (HCC) from metastatic carcinomas to the liver is often difficult, if not impossible, particularly in needle biopsy and fine-needle aspiration specimens. In an attempt to identify a specific immunohistochemical profile that would distinguish HCC from metastatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service