Skip to Content
Merck
All Photos(1)

Key Documents

21627-M

Millipore

IHC Select Proteinase K

Synonym(s):

Antigen/ Epitope Retrieval Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053

packaging

pkg of 15 mL

Quality Level

manufacturer/tradename

Chemicon®
IHC Select

technique(s)

immunohistochemistry: suitable (paraffin)

Application

For use as a Pretreatment Reagent in Immunohistochemical staining procedures. Use Proteinase K to retrieve Epitopes in formalin-fixed, paraffin-embedded tissue sections mounted on glass microscope slides. This procedure is performed after deparaffinization and prior to IHC.



Warm to room temperature before use. Digestion time and dilution should be determined by user as concentration is dependent on the type of antibody, antigen, and fixation method used. Recommended starting dilution range: Neat to 1:30.



For best results use this reagent in conjunction with other IHC Select detection reagents. A complete list of reagents is available at www.chemicon.com.

Physical form

Liquid Proteinase K containing 15mM sodium azide as a preservative. Discard if it becomes cloudy or contaminated.

Storage and Stability

When stored at 2-8°C, the Proteinase K is stable up to the expiration date printed on the label. Do not freeze or expose to elevated temperatures.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aisan Farhadi et al.
The European journal of neuroscience, 56(1), 3755-3778 (2022-05-06)
Diabetes mellitus may cause tau protein hyperphosphorylation and neurodegeneration, but the exact mechanism by which diabetic conditions induce tau pathology remains unclear. Tau protein hyperphosphorylation is considered a major pathological hallmark of neurodegeneration and can be triggered by diabetes. Various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service