GC Analysis of Glycidol Enantiomers on Astec® CHIRALDEX™ B-TA
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Astec CHIRALDEX B-TA, 30 m x 0.25 mm I.D., 0.12 μm (73023AST)
oven
80 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peak 1: S(-)-glycidol
peak 2: R(+)-glycidol
Description
Analysis Note
Elution order reversal on CHIRALDEX A-TA and CHIRALDEX G-TA.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC