Merck
  • The transcription factor Ikaros inhibits cell proliferation by downregulating ANXA4 expression in hepatocellular carcinoma.
The transcription factor Ikaros inhibits cell proliferation by downregulating ANXA4 expression in hepatocellular carcinoma.
American journal of cancer research (2017-07-04)
Yi-Yao Liu, Chao Ge, Hua Tian, Jing-Yi Jiang, Fang-Yu Zhao, Hong Li, Tao-Yang Chen, Ming Yao, Jin-Jun Li
ABSTRACT
The occurrence and progression of hepatocellular carcinoma (HCC) are affected by complicated signal transduction factors. Our previous study identified Ikaros as a novel reactivated therapeutic target that acts as a transcriptional repressor and reactivates anticancer mechanisms in HCC therapy. Annexin A4 (ANXA4) is a member of the Annexin family that plays an essential role in several cancers, but it has not been investigated in HCC proliferation. Using cDNA microarrays, ANXA4 was shown to be associated with Ikaros in Ikaros-overexpressing cells. The aim of this work was to characterize the relationship between Ikaros and ANXA4 and the role of ANXA4 in HCC. The effect of Ikaros on ANXA4 was analyzed in HCC cell lines and HCC patient samples, and functional recovery experiments were performed between Ikaros and ANXA4. Furthermore, the effect of ANXA4 on cell proliferation
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ANXA4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution