Merck
Calcium and electrical dynamics in lymphatic endothelium.
The Journal of physiology (2017-10-11)
Erik J Behringer, Joshua P Scallan, Mohammad Jafarnejad, Jorge A Castorena-Gonzalez, Scott D Zawieja, James E Moore, Michael J Davis, Steven S Segal
ABSTRACT
Endothelial cell function in resistance arteries integrates Ca Subsequent to a rise in intracellular Ca
Sigma-Aldrich
NS309, ≥98% (HPLC), solid