Merck
  • The measurement of bisphenol A and its analogues, perfluorinated compounds in twenty species of freshwater and marine fishes, a time-trend comparison and human health based assessment.
The measurement of bisphenol A and its analogues, perfluorinated compounds in twenty species of freshwater and marine fishes, a time-trend comparison and human health based assessment.
Marine pollution bulletin (2017-05-30)
Y M Wong, R Li, C K F Lee, H T Wan, Chris K C Wong
ABSTRACT
Our previous study in 2011 reported the detection of BPA and PFAAs in 20 species of marine and freshwater fishes. With an emerging evidence to suggest the metabolic-disrupting effects of BPA/PFAAs in animals, the present study was aimed to provide a time-trend analysis to determine the current concentrations of PFAAs and BPA in 20 commercially available Hong Kong species of fishes. Since the manufacture and use of BPA is being prohibited in most nations, the introduction of BPA alternatives has recently been incorporated in the markets. Therefore, the concentrations of BPB, BPF and BPS were determined. In the present study, all freshwater and seawater fish samples showed quantified concentrations [>Limit of Quantification (LOQ<0.5ng/g)] of BPA. BPF was detected in some marine (yellow seafin, bigeye, goldspotted rabbitfish, snubnose pompano, tongue sole, Bleeker's grouper and orange-spotted grouper) and freshwater fishes (mud carp, crucian carp, tilapia, catfish, mandarin fish, grass carp, grey mullet and spotted snakehead). Two of the compounds, BPS and BPB could only be identified in the marine fishes (snubnose pompano, yellow seafin). In PFAA analysis, PFOA, PFDA, PFOS, PFUdA and PFDoA were found in most of the marine and freshwater fishes. PFOS and PFOA were shown to be the two predominant PFAAs in fishes. On the basis of the measured concentrations of bisphenols, BPs (BPA, BPB, BPF, BPS) and PFAAs, the average daily intake for BPs (20.5-31.5ng/kgb.w./day) and PFAAs (1.17-1.83ng/kgb.w./day) were calculated and found to be lower than values of tolerable daily intake (TDI) established in Europe. However, as compared with our previous study in 2011, the present study revealed an approximate 10-fold increase in the concentrations of BPA in the fish samples. Although the hazard ratio of consuming fishes for BPA and PFAA exposure is expected to remain low, possible additive metabolic-disrupting effect of BPA and its analogues as well PFAAs should be taken into consideration for human health risk assessment.
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%
Sigma-Aldrich
Bis(4-hydroxyphenyl)methane, 98%