Merck
  • Soft, Wet-Chemical Synthesis of Metastable Superparamagnetic Hexagonal Close-Packed Nickel Nanoparticles in Different Ionic Liquids.
Soft, Wet-Chemical Synthesis of Metastable Superparamagnetic Hexagonal Close-Packed Nickel Nanoparticles in Different Ionic Liquids.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-02-15)
Susann Wegner, Christina Rutz, Kai Schütte, Juri Barthel, Alexey Bushmelev, Annette Schmidt, Katharina Dilchert, Roland A Fischer, Christoph Janiak
ABSTRACT
The microwave-induced decomposition of bis{N,N'-diisopropylacetamidinate}nickel(II) [Ni{MeC(NiPr)
Sigma-Aldrich
1-Chlorooctane, 99%
Sigma-Aldrich
DIC, 99%
Sigma-Aldrich
Bis(1,5-cyclooctadiene)nickel(0)
Sigma-Aldrich
Diisopropylcarbodiimide solution, 1 M in THF