Merck
MicroRNA-126 regulates HOXA9 by binding to the homeobox.
Molecular and cellular biology (2008-05-14)
Wei-Fang Shen, Yu-Long Hu, Lalita Uttarwar, Emmanuelle Passegue, Corey Largman
ABSTRACT
The PicTar program predicted that microRNA-126 (miR-126), miR-145, and let-7s target highly conserved sites within the Hoxa9 homeobox. There are increased nucleotide constraints in the three microRNA seed sites among Hoxa9 genes beyond that required to maintain protein identity, suggesting additional functional conservation. In preliminary experiments, forced expression of these microRNAs in Hoxa9-immortalized bone marrow cells downregulated the HOXA9 protein and caused loss of biological activity. The microRNAs were shown to target their predicted sites within the homeobox. miR-126 and Hoxa9 mRNA are coexpressed in hematopoietic stem cells and downregulated in parallel during progenitor cell differentiation; however, miR-145 is barely detectable in hematopoietic cells, and let-7s are highly expressed in bone marrow progenitors, suggesting that miR-126 may function in normal hematopoietic cells to modulate HOXA9 protein. In support of this hypothesis, expression of miR-126 alone in MLL-ENL-immortalized bone marrow cells decreased endogenous HOXA9 protein, while inhibition of endogenous miR-126 increased expression of HOXA9 in F9 cells.