Merck
  • Discovering inhibitors of human sirtuin type 2: novel structural scaffolds.
Discovering inhibitors of human sirtuin type 2: novel structural scaffolds.
Journal of medicinal chemistry (2006-11-28)
Anu J Tervo, Tiina Suuronen, Sergiy Kyrylenko, Erkki Kuusisto, Päivi H Kiviranta, Antero Salminen, Jukka Leppänen, Antti Poso
ABSTRACT
A successful virtual screening experiment of novel SIRT2 inhibitors is described. Four out of 11 experimentally tested compounds showed in vitro inhibitory activity toward SIRT2 in a micromolar level, resulting in an experimental hit ratio of 36%. Two of these compounds inhibited SIRT2 with IC50 (microM) values of 51 and 91; moreover, one of the new inhibitors was comprised of an entirely new SIRT2-inhibiting structural scaffold.
Sigma-Aldrich
JFD00244, ≥98% (HPLC), solid