Merck
  • SHMT2 drives glioma cell survival in ischaemia but imposes a dependence on glycine clearance.
SHMT2 drives glioma cell survival in ischaemia but imposes a dependence on glycine clearance.
Nature (2015-04-10)
Dohoon Kim, Brian P Fiske, Kivanc Birsoy, Elizaveta Freinkman, Kenjiro Kami, Richard L Possemato, Yakov Chudnovsky, Michael E Pacold, Walter W Chen, Jason R Cantor, Laura M Shelton, Dan Y Gui, Manjae Kwon, Shakti H Ramkissoon, Keith L Ligon, Seong Woo Kang, Matija Snuderl, Matthew G Vander Heiden, David M Sabatini
ABSTRACT
Cancer cells adapt their metabolic processes to support rapid proliferation, but less is known about how cancer cells alter metabolism to promote cell survival in a poorly vascularized tumour microenvironment. Here we identify a key role for serine and glycine metabolism in the survival of brain cancer cells within the ischaemic zones of gliomas. In human glioblastoma multiforme, mitochondrial serine hydroxymethyltransferase (SHMT2) and glycine decarboxylase (GLDC) are highly expressed in the pseudopalisading cells that surround necrotic foci. We find that SHMT2 activity limits that of pyruvate kinase (PKM2) and reduces oxygen consumption, eliciting a metabolic state that confers a profound survival advantage to cells in poorly vascularized tumour regions. GLDC inhibition impairs cells with high SHMT2 levels as the excess glycine not metabolized by GLDC can be converted to the toxic molecules aminoacetone and methylglyoxal. Thus, SHMT2 is required for cancer cells to adapt to the tumour environment, but also renders these cells sensitive to glycine cleavage system inhibition.
Sigma-Aldrich
Anti-SHMT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-GLDC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution