Merck
  • Continuous colorimetric screening assay for detection of d-amino acid aminotransferase mutants displaying altered substrate specificity.
Continuous colorimetric screening assay for detection of d-amino acid aminotransferase mutants displaying altered substrate specificity.
Analytical biochemistry (2014-06-21)
Janet E B Barber, Adam M Damry, Guido F Calderini, Curtis J W Walton, Roberto A Chica
ABSTRACT
D-Amino acid aminotransferase (DAAT) catalyzes the synthesis of numerous d-amino acids, making it an attractive biocatalyst for the production of enantiopure d-amino acids. To bolster its biocatalytic applicability, improved variants displaying increased activity toward non-native substrates are desired. Here, we report the development of a high-throughput, colorimetric, continuous coupled enzyme assay for the screening of DAAT mutant libraries that is based on the use of d-amino acid oxidase (DAAO). In this assay, the d-amino acid product of DAAT is oxidized by DAAO with concomitant release of hydrogen peroxide, which is detected colorimetrically by the addition of horseradish peroxidase and o-dianisidine. Using this assay, we measured apparent KM and kcat values for DAAT and identified mutants displaying altered substrate specificity via the screening of cell lysates in 96-well plates. The DAAO coupled assay is sensitive in that it allowed the detection of a DAAT mutant displaying an approximately 2000-fold decrease in kcat/KM relative to wild type. In addition, the DAAO assay enabled the identification of two DAAT mutants (V33Y and V33G) that are more efficient than wild type at transaminating the non-native acceptor phenylpyruvate. We expect that this assay will be useful for the engineering of additional mutants displaying increased activity toward non-native substrates.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Potassium phosphate tribasic, Vetec, reagent grade, 97%
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Supelco
Nitrilotriacetic acid, Pharmaceutical Secondary Standard; Certified Reference Material