Merck
  • Enzymatic enantiomeric resolution of phenylethylamines structurally related to amphetamine.
Enzymatic enantiomeric resolution of phenylethylamines structurally related to amphetamine.
Organic & biomolecular chemistry (2011-10-06)
Lourdes Muñoz, Anna M Rodriguez, Gloria Rosell, M Pilar Bosch, Angel Guerrero
ABSTRACT
Both enantiomers of several phenylethylamines, structurally related to amphetamine, have been prepared in good yields and excellent enantiomeric purity by enzymatic kinetic resolution using CAL-B and ethyl methoxyacetate as the acyl donor. In the case of the 4-hydroxyderivative of amphetamine (compound 4i), the S enantiomer racemized possibly in a dynamic kinetic resolution (DKR) under the enzymatic conditions used.
Sigma-Aldrich
Ethyl methoxyacetate, 97%
Sign Into View Organizational & Contract Pricing