Merck
Interaction of transketolase from human tissues with substrates.
Biochemistry. Biokhimiia (2011-11-16)
L E Meshalkina, O N Solovjeva, G A Kochetov
ABSTRACT
The Michaelis constant values for substrates of transketolase from human tissues were determined over a wide range of substrate concentrations. It is shown that K(m) values determined by other authors are significantly overestimated and explained why this is so.
Sigma-Aldrich
D-Ribose 5-phosphate barium salt hexahydrate, ≥99.0% (TLC)