Merck
  • Urinary 2-methoxy acetic acid accumulation in response to 2-methoxy ethanol exposure.
Urinary 2-methoxy acetic acid accumulation in response to 2-methoxy ethanol exposure.
Archives of environmental health (2001-03-21)
T S Shih, S H Liou, C Y Chen, T J Smith
ABSTRACT
Urinary 2-methoxy acetic acid reportedly has a long half-life (77.1 hr) in humans. The authors studied urinary 2-methoxy acetic acid in a group of 18 workers exposed to 2-methoxy ethanol from Monday to Saturday following a 7-d cease in production. The weekly time-weighted average exposure concentration of 2-methoxy ethanol was 4.5 ppm. The urinary 2-methoxy acetic acid of exposed workers was increased significantly, from 18.5 microg/ml (10.6 mg/gm creatinine) on Monday (prior to work) to 48.4 microg/ml (46.5 mg/gm creatinine) on Friday (after work), to 51.2 microg/ml (45.6 mg/gm creatinine) on Saturday after work. These levels occurred, despite that fact that the daily mean time-weighted average 2-methoxy ethanol exposures were very consistent and were close to the current Taiwan Permissible Exposure Limit of 5 ppm. These urinary 2-methoxy acetic acid levels were much higher than levels that occur with inhalation only, and they demonstrate that skin absorption is a significant contributor to 2-methoxy ethanol exposure. The high background concentrations of 2-methoxy acetic acid in the preshift urine samples following a 7-d production halt confirm that there is a long half-life of 2-methoxy acetic acid in humans. The determination of urinary 2-methoxy acetic acid is recommended for exposure assessment of 2-methoxy ethanol.
Sigma-Aldrich
Ethyl methoxyacetate, 97%
Pricing and availability is not currently available. Contact Technical Service