Merck
  • Influence of postpartum supplementation with vitamin A on the levels of immunoglobulin A in human colostrum.
Influence of postpartum supplementation with vitamin A on the levels of immunoglobulin A in human colostrum.
Jornal de pediatria (2012-03-13)
Mayara S R Lima, Penha P C Ribeiro, Jovilma M S Medeiros, Isabelle F Silva, Ana C P Medeiros, Roberto Dimenstein
ABSTRACT
To evaluate the influence of supplementation with retinyl palmitate in the immediate postpartum period on the levels of secretory immunoglobulin A (SIgA) in colostrum. A clinical trial was conducted among 96 recently-delivered mothers treated at a public maternity hospital, divided into control group, which was not supplemented (n = 44), and test group, supplemented on the first day postpartum (n = 52). Samples of 2 mL of colostrum were collected on the first 2 days postpartum. SIgA was measured by turbidimetry and data were analyzed by the Student t test. Before supplementation, the average SIgA was 829.1±337.6 mg/dL in the control group and 827.3±249.8 mg/dL in the test group (p = 0.52). After supplementation, the average SIgA was 343.9±177.2 mg/dL in the unsupplemented group and 501.2±54.5 mg/dL in the supplemented group (p < 0.00001). The colostrum of women supplemented with retinyl palmitate has higher levels of SIgA, which suggests that the production of antibodies is modulated by vitamin A.
Product Description
Supelco
Retinol palmitate, analytical standard
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g