Merck
  • Near infrared fluorescence lymphatic laparoscopy of the colon and mesocolon.
Near infrared fluorescence lymphatic laparoscopy of the colon and mesocolon.
Colorectal disease : the official journal of the Association of Coloproctology of Great Britain and Ireland (2011-12-07)
M Ankersmit, M H G M van der Pas, D A van Dam, W J H J Meijerink
ABSTRACT
During surgery, a surgeon relies on the vision of his eyes and the touch of his hands. While laparoscopic surgery for colon cancer has proven to be safe and effective, it still remains a technically difficult procedure. Although it is associated with reduced haptic feedback, by enforcing the power of visual guidance, the loss of this feedback can be (partly) compensated for. Here we describe how the use of near-infrared dyes and fluorescence laparoscopy could help improve tumour staging and therefore lead to better selection of patients for postoperative adjuvant chemotherapy. More controversially, and analogous to melanoma and breast cancer surgery with sentinel node biopsy, we speculate that local resection with SLN harvesting in early colon cancer might change the therapeutic and surgical strategy in colon cancer.
Sigma-Aldrich
Patent Blue VF, Dye content 50 %
Sigma-Aldrich
Patent blue VF sodium salt, for microscopy